siRNA Details
| Gene Symbol: ZNF786 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152411.3 | NP_689624.2 | 4 | 2541 | ||
| GenBank mRNA | AK095701.1 | - | 3 | 2384 | ||
| AK297472.1 | - | 5 | 2805 | |||
| AL834510.1 | - | 4 | 2459 | |||
| BX647929.1 | - | 3 | 2379 | |||

ZNF786
Genomic Map