siRNA Details
| Gene Symbol: ZNF746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163474.1 | NP_001156946.1 | 7 | 2926 | ||
| NM_152557.4 | NP_689770.3 | 7 | 2923 | |||
| XM_005249955.2 | XP_005250012.1 | 7 | 2996 | |||
| XM_005249956.2 | XP_005250013.1 | 7 | 2993 | |||
| XM_005249957.1 | XP_005250014.1 | 7 | 3098 | |||
| XM_011515838.1 | XP_011514140.1 | 7 | 3053 | |||
| GenBank mRNA | AK055975.1 | - | 1 | 1822 | ||
| AK128244.1 | - | 5 | 2283 | |||
| BC031968.1 | - | 1 | 521 | |||
| BC068505.1 | - | 7 | 2819 | 30347892 | ||

ZNF746
Genomic Map