siRNA Details
| Gene Symbol: LINGO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258282.1 | NP_001245211.1 | 7 | 1449 | ||
| NM_152570.2 | NP_689783.1 | 7 | 1529 | |||
| XM_011517719.1 | XP_011516021.1 | 6 | 2397 | |||
| XM_011517724.2 | XP_011516026.1 | 5 | 5416 | |||
| XM_011517728.2 | XP_011516030.1 | 2 | 5195 | |||
| XM_017014303.1 | XP_016869792.1 | 11 | 5300 | |||
| XM_017014304.1 | XP_016869793.1 | 4 | 7820 | |||
| XM_017014305.1 | XP_016869794.1 | 5 | 7403 | |||
| XM_017014306.1 | XP_016869795.1 | 3 | 4564 | |||
| XM_017014307.1 | XP_016869796.1 | 2 | 2037 | |||
| GenBank mRNA | AK056372.1 | - | 7 | 1529 | ||
| AK172834.1 | - | 1 | 529 | |||
| AK290972.1 | - | 2 | 1060 | |||
| AY358114.1 | - | 2 | 1083 | |||
| BC137514.1 | - | 7 | 1389 | 9021514 | ||
| BC144676.1 | - | 7 | 1389 | 9053210 | ||

LINGO2
Genomic Map