siRNA Details
| Gene Symbol: OIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152635.2 | NP_689848.1 | 5 | 1022 | ||
| NR_130125.1 | 5 | 1022 | ||||
| GenBank mRNA | AK096435.1 | - | 5 | 997 | ||
| AY013707.2 | - | 5 | 853 | |||
| AY358339.1 | - | 5 | 975 | |||
| BC126440.1 | - | 5 | 994 | 8992156 | ||

OIT3
Genomic Map