siRNA Details
| Gene Symbol: SLC39A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145195.1 | NP_001138667.1 | 3 | 796 | ||
| NM_001282733.1 | NP_001269662.1 | 3 | 796 | |||
| NM_001282734.1 | NP_001269663.1 | 2 | 449 | |||
| NM_152725.3 | NP_689938.2 | 3 | 796 | |||
| GenBank mRNA | AK055061.1 | - | 3 | 744 | ||
| AK295294.1 | - | 2 | 325 | |||
| AK303735.1 | - | 3 | 481 | |||
| BC035118.1 | - | 3 | 728 | |||
| BC047635.1 | - | 3 | 711 | 4820920 | ||
| BC065917.1 | - | 2 | 540 | |||
| BC094700.1 | - | 3 | 672 | 30407123 | ||
| BC117323.1 | - | 2 | 546 | 40125874 | ||
| BC143551.1 | - | 2 | 545 | 9052062 | ||

SLC39A12
Genomic Map