siRNA Details
| Gene Symbol: UHRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152896.2 | NP_690856.1 | 2 | 612 | ||
| NR_046386.1 | 2 | 612 | ||||
| XM_011517704.2 | XP_011516006.1 | 2 | 241 | |||
| XM_017014253.1 | XP_016869742.1 | 2 | 615 | |||
| XM_017014254.1 | XP_016869743.1 | 2 | 241 | |||
| GenBank mRNA | AB071698.1 | - | 2 | 310 | ||
| AF274049.1 | - | 2 | 440 | |||
| AK308433.1 | - | 2 | 564 | |||
| BC028397.2 | - | 2 | 612 | 4797830 | ||
| BG709183.1 | - | 2 | 618 | |||

UHRF2
Genomic Map