siRNA Details
| Gene Symbol: UHRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152896.2 | NP_690856.1 | 2 | 659 | ||
| NR_046386.1 | 2 | 659 | ||||
| XM_011517704.2 | XP_011516006.1 | 2 | 288 | |||
| XM_011517705.2 | XP_011516007.1 | 1 | 47 | |||
| XM_017014253.1 | XP_016869742.1 | 2 | 662 | |||
| XM_017014254.1 | XP_016869743.1 | 2 | 288 | |||
| GenBank mRNA | AB071698.1 | - | 2 | 357 | ||
| AF274049.1 | - | 2 | 487 | |||
| AK308433.1 | - | 2 | 611 | |||
| BC028397.2 | - | 2 | 659 | 4797830 | ||
| BG709183.1 | - | 2 | 665 | |||

UHRF2
Genomic Map