siRNA Details
| Gene Symbol: NFXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278623.1 | NP_001265552.1 | 7 | 977 | ||
| NM_001278624.1 | NP_001265553.1 | 7 | 1115 | |||
| NM_152995.5 | NP_694540.3 | 7 | 1017 | |||
| NR_103795.1 | 7 | 977 | ||||
| GenBank mRNA | AB085695.1 | - | 7 | 973 | ||
| AB181916.1 | - | 7 | 1097 | |||
| AK131285.1 | - | 5 | 563 | |||
| AK302440.1 | - | 7 | 975 | |||
| BC051193.1 | - | 7 | 948 | 5271401 | ||

NFXL1
Genomic Map