siRNA Details
| Gene Symbol: NFXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278623.1 | NP_001265552.1 | 23 | 3534 | ||
| NM_001278624.1 | NP_001265553.1 | 23 | 3672 | |||
| NM_152995.5 | NP_694540.3 | 23 | 3574 | |||
| NR_103795.1 | 24 | 3714 | ||||
| GenBank mRNA | AB085695.1 | - | 23 | 3528 | ||
| AY134856.1 | - | 7 | 1577 | |||
| BC018019.1 | - | 6 | 1267 | |||
| BC051193.1 | - | 24 | 3684 | 5271401 | ||
| BM724120.1 | - | 1 | 696 | |||

NFXL1
Genomic Map