siRNA Details
| Gene Symbol: POU5F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153216.1 | NP_694948.1 | 1 | 1094 | ||
| GenBank mRNA | AK098546.1 | - | 1 | 1094 | ||
| BC029532.1 | - | 1 | 1054 | 5295125 | ||
| BX641019.1 | - | 1 | 614 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

POU5F2| Gene Symbol: POU5F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153216.1 | NP_694948.1 | 1 | 1094 | ||
| GenBank mRNA | AK098546.1 | - | 1 | 1094 | ||
| BC029532.1 | - | 1 | 1054 | 5295125 | ||
| BX641019.1 | - | 1 | 614 | |||