siRNA Details
| Gene Symbol: ZNF550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277090.1 | NP_001264019.1 | 5 | 2157 | ||
| NM_001277091.1 | NP_001264020.1 | 5 | 2157 | |||
| XM_011526567.2 | XP_011524869.1 | 5 | 2154 | |||
| XM_011526568.2 | XP_011524870.1 | 5 | 3247 | |||
| GenBank mRNA | AL833214.1 | - | 2 | 1726 | ||
| BC034810.1 | - | 5 | 2133 | 4823435 | ||
| BC053858.1 | - | 5 | 2131 | 6044705 | ||

ZNF550
Genomic Map