siRNA Details
| Gene Symbol: LRRC57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153260.2 | NP_694992.2 | 6 | 891 | ||
| GenBank mRNA | AK094131.1 | - | 6 | 885 | ||
| AK094891.1 | - | 5 | 1125 | |||
| BC058935.1 | - | 6 | 847 | 5578024 | ||
| BX538324.1 | - | 5 | 1037 | |||
| CN350663.1 | - | 3 | 341 | |||
| CR749223.1 | - | 6 | 860 | |||

LRRC57
Genomic Map