siRNA Details
| Gene Symbol: IFNLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170743.3 | NP_734464.1 | 7 | 2136 | ||
| NM_173064.2 | NP_775087.1 | 7 | 2049 | |||
| NM_173065.2 | NP_775088.1 | 6 | 2005 | |||
| XM_006710394.3 | XP_006710457.2 | 6 | 2200 | |||
| XM_017000479.1 | XP_016855968.1 | 6 | 2165 | |||
| GenBank mRNA | AF439325.1 | - | 7 | 2109 | ||
| AK160364.1 | - | 7 | 2112 | |||
| BC140872.1 | - | 7 | 2185 | 9021740 | ||

IFNLR1
Genomic Map