siRNA Details
| Gene Symbol: TPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173353.3 | NP_775489.2 | 2, 3 | 404 | ||
| GenBank mRNA | AK094614.1 | - | 2, 3 | 273 | ||
| AY098914.1 | - | 2, 3 | 404 | |||
| BC114442.1 | - | 2, 3 | 359 | 40083878 | ||
| BC114499.1 | - | 2, 3 | 341 | 40083876 | ||

TPH2
Genomic Map