siRNA Details
Gene Symbol: RNF149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173647.3 | NP_775918.2 | 7 | 2073 | ||
GenBank mRNA | AK074985.1 | - | 7 | 2047 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RNF149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173647.3 | NP_775918.2 | 7 | 2073 | ||
GenBank mRNA | AK074985.1 | - | 7 | 2047 |