siRNA Details
| Gene Symbol: ZNF800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176814.4 | NP_789784.2 | 6 | 3627 | ||
| XM_005250181.3 | XP_005250238.1 | 6 | 3701 | |||
| GenBank mRNA | AK125291.1 | - | 2 | 1431 | ||
| BC037348.1 | - | 2 | 2601 | |||
| BC110623.1 | - | 6 | 3172 | 40036036 | ||
| BC110624.1 | - | 6 | 3167 | 40036038 | ||

ZNF800
Genomic Map