siRNA Details
| Gene Symbol: ZNF800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176814.4 | NP_789784.2 | 6 | 3937 | ||
| XM_005250181.3 | XP_005250238.1 | 6 | 4011 | |||
| GenBank mRNA | AK125291.1 | - | 2 | 1741 | ||
| BC037348.1 | - | 2 | 2911 | |||
| BG112338.1 | - | 1 | 33 | |||

ZNF800
Genomic Map