siRNA Details
| Gene Symbol: DHFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195643.1 | NP_001182572.1 | 2 | 1652 | ||
| NM_176815.4 | NP_789785.1 | 2 | 1539 | |||
| XM_011512537.2 | XP_011510839.1 | 2 | 1717 | |||
| GenBank mRNA | AK122674.1 | - | 2 | 1652 | ||
| AL832912.1 | - | 2 | 1328 | |||
| BC045541.1 | - | 2 | 1370 | 5262102 | ||
| BC063379.1 | - | 2 | 1328 | 4109721 | ||

DHFR2
Genomic Map