siRNA Details
| Gene Symbol: CYP4X1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320289.1 | NP_001307218.1 | 7 | 1049 | ||
| NM_001320290.1 | NP_001307219.1 | 7 | 786 | |||
| NM_178033.1 | NP_828847.1 | 7 | 1083 | |||
| XM_017000973.1 | XP_016856462.1 | 7 | 1114 | |||
| GenBank mRNA | AK091806.1 | - | 7 | 787 | ||
| AK098065.1 | - | 7 | 1076 | |||
| AK131355.1 | - | 7 | 1049 | |||
| AM040940.1 | - | 7 | 920 | |||
| AY358537.1 | - | 7 | 1066 | |||
| BC028102.1 | - | 7 | 1083 | 5243005 | ||

CYP4X1
Genomic Map