siRNA Details
| Gene Symbol: CYP4X1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320289.1 | NP_001307218.1 | 12 | 1619 | ||
| NM_001320290.1 | NP_001307219.1 | 12 | 1356 | |||
| NM_178033.1 | NP_828847.1 | 12 | 1653 | |||
| GenBank mRNA | AK091806.1 | - | 12 | 1357 | ||
| AK098065.1 | - | 12 | 1646 | |||
| AK131355.1 | - | 12 | 1619 | |||
| AM040940.1 | - | 12 | 1490 | |||
| AY358537.1 | - | 12 | 1636 | |||
| BC028102.1 | - | 12 | 1653 | 5243005 | ||

CYP4X1
Genomic Map