siRNA Details
| Gene Symbol: HSD17B13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136230.2 | NP_001129702.1 | 6 | 1525 | ||
| NM_178135.4 | NP_835236.2 | 7 | 1633 | |||
| GenBank mRNA | AB073347.1 | - | 7 | 1602 | ||
| AY186249.1 | - | 7 | 1613 | |||
| AY186250.1 | - | 6 | 1505 | |||
| AY358575.1 | - | 7 | 1633 | |||

HSD17B13
Genomic Map