siRNA Details
| Gene Symbol: SLC5A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049207.1 | - | 9 | 1861 | 5188979 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC5A12| Gene Symbol: SLC5A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC049207.1 | - | 9 | 1861 | 5188979 | |