siRNA Details
| Gene Symbol: ZNF546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297763.1 | NP_001284692.1 | 6 | 554 | ||
| NM_178544.4 | NP_848639.2 | 6 | 632 | |||
| XM_011526898.2 | XP_011525200.1 | 5 | 1699 | |||
| XM_011526899.2 | XP_011525201.1 | 6 | 544 | |||
| GenBank mRNA | AK122968.1 | - | 6 | 526 | ||
| AK292528.1 | - | 6 | 580 | |||
| BC045649.1 | - | 6 | 546 | 5268795 | ||
| DA217689.1 | - | 5 | 469 | |||

ZNF546
Genomic Map