siRNA Details
| Gene Symbol: ZNF546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297763.1 | NP_001284692.1 | 7 | 2676 | ||
| NM_178544.4 | NP_848639.2 | 7 | 2754 | |||
| XM_011526898.2 | XP_011525200.1 | 6 | 3821 | |||
| XM_011526899.2 | XP_011525201.1 | 7 | 2666 | |||
| GenBank mRNA | AK122968.1 | - | 7 | 2648 | ||
| AK292528.1 | - | 7 | 2702 | |||
| BC045649.1 | - | 7 | 2668 | 5268795 | ||

ZNF546
Genomic Map