siRNA Details
| Gene Symbol: TRIML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178556.3 | NP_848651.2 | 3 | 758 | ||
| XM_006714207.3 | XP_006714270.1 | 3 | 783 | |||
| XM_011531919.1 | XP_011530221.1 | 4 | 610 | |||
| GenBank mRNA | AK093499.1 | - | 3 | 758 | ||
| BC113860.1 | - | 3 | 711 | 40033174 | ||
| BC114469.1 | - | 3 | 711 | 40033173 | ||

TRIML1
Genomic Map