siRNA Details
| Gene Symbol: GPR180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180989.5 | NP_851320.1 | 9 | 2016 | ||
| GenBank mRNA | AK024059.1 | - | 1 | 444 | ||
| AK126794.1 | - | 9 | 1921 | |||
| BC033906.1 | - | 8 | 1810 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR180| Gene Symbol: GPR180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_180989.5 | NP_851320.1 | 9 | 2016 | ||
| GenBank mRNA | AK024059.1 | - | 1 | 444 | ||
| AK126794.1 | - | 9 | 1921 | |||
| BC033906.1 | - | 8 | 1810 | |||