siRNA Details
| Gene Symbol: GRASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271856.1 | NP_001258785.1 | 4, 5 | 384 | ||
| NM_181711.3 | NP_859062.1 | 5, 6 | 618 | |||
| XM_005268691.3 | XP_005268748.1 | 5, 6 | 1786 | |||
| XM_011537996.1 | XP_011536298.1 | 4, 5 | 1564 | |||
| GenBank mRNA | BC035500.2 | - | 4, 5 | 355 | 4422992 | |
| BC052967.1 | - | 4, 5 | 302 | |||
| BC053666.1 | - | 5, 6 | 599 | 6151168 | ||
| HY035281.1 | - | 4, 5 | 318 | |||

GRASP
Genomic Map