siRNA Details
| Gene Symbol: NCOA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122842.2 | NP_001116314.1 | 3 | 608 | ||
| NM_001199619.1 | NP_001186548.1 | 4 | 591 | |||
| NM_001199620.1 | NP_001186549.1 | 5 | 709 | |||
| NM_181782.4 | NP_861447.3 | 3 | 608 | |||
| XM_005266822.4 | XP_005266879.1 | 3 | 638 | |||
| XM_006715340.3 | XP_006715403.1 | 3 | 565 | |||
| XM_011535455.2 | XP_011533757.1 | 3 | 513 | |||
| XM_017010269.1 | XP_016865758.1 | 3 | 324 | |||
| XM_017010270.1 | XP_016865759.1 | 4 | 570 | |||
| XM_017010271.1 | XP_016865760.1 | 3 | 568 | |||
| XM_017010272.1 | XP_016865761.1 | 3 | 421 | |||
| XM_017010273.1 | XP_016865762.1 | 3 | 638 | |||
| XM_017010274.1 | XP_016865763.1 | 3 | 727 | |||
| GenBank mRNA | AB074157.1 | - | 3 | 408 | ||
| AF493978.1 | - | 4 | 591 | |||
| AK127512.1 | - | 3 | 418 | |||
| AK307395.1 | - | 4 | 570 | |||
| AL832621.2 | - | 3 | 401 | |||
| AL832628.2 | - | 3 | 365 | |||
| AL834442.1 | - | 1 | 29 | |||
| BC036461.1 | - | 3 | 436 | |||
| BC071782.1 | - | 2 | 376 | |||
| BC137094.1 | - | 5 | 625 | 9021091 | ||
| BC137095.1 | - | 4 | 507 | 9021092 | ||
| BX537385.1 | - | 3 | 403 | |||

NCOA7
Genomic Map