siRNA Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098475.1 | NP_001091945.1 | 12 | 2223 | ||
| NM_182499.3 | NP_872305.3 | 13 | 2025 | |||
| XM_006711156.2 | XP_006711219.1 | 7 | 1853 | |||
| XM_011509152.1 | XP_011507454.1 | 13 | 1612 | |||
| XM_011509153.1 | XP_011507455.1 | 14 | 1503 | |||
| XM_011509154.1 | XP_011507456.1 | 13 | 1570 | |||
| XM_011509155.1 | XP_011507457.1 | 14 | 1414 | |||
| XM_011509156.1 | XP_011507458.1 | 12 | 1537 | |||
| XM_011509157.1 | XP_011507459.1 | 13 | 1339 | |||
| XM_011509158.2 | XP_011507460.1 | 8 | 1327 | |||
| XM_011509159.2 | XP_011507461.1 | 8 | 1318 | |||
| XM_011509160.2 | XP_011507462.1 | 8 | 1363 | |||
| XM_011509161.1 | XP_011507463.1 | 8 | 1331 | |||
| XM_017000255.1 | XP_016855744.1 | 7 | 1257 | |||
| GenBank mRNA | AL713777.1 | - | 8 | 1018 | ||
| AY358195.1 | - | 6 | 992 | |||
| BC044864.1 | - | 13 | 2024 | 4836747 | ||

TDRD10
Genomic Map