siRNA Details
| Gene Symbol: ZNF605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164715.1 | NP_001158187.1 | 5 | 2790 | ||
| NM_183238.3 | NP_899061.1 | 5 | 2796 | |||
| GenBank mRNA | AK122878.1 | - | 5 | 2790 | ||
| AL832191.1 | - | 5 | 2797 | |||
| AL832623.2 | - | 5 | 2821 | |||

ZNF605
Genomic Map