siRNA Details
| Gene Symbol: GPR151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194251.2 | NP_919227.2 | 1 | 864 | ||
| GenBank mRNA | AY351676.1 | - | 1 | 864 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPR151| Gene Symbol: GPR151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194251.2 | NP_919227.2 | 1 | 864 | ||
| GenBank mRNA | AY351676.1 | - | 1 | 864 | ||
Set Membership: |
|