siRNA Details
Gene Symbol: NAIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197956.3 | NP_931045.1 | 2 | 1785 | ||
GenBank mRNA | AK122729.1 | - | 2 | 1785 | ||
AL833392.1 | - | 2 | 1577 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NAIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_197956.3 | NP_931045.1 | 2 | 1785 | ||
GenBank mRNA | AK122729.1 | - | 2 | 1785 | ||
AL833392.1 | - | 2 | 1577 |