siRNA Details
| Gene Symbol: AGXT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000030.2 | NP_000021.1 | 2 | 312 | ||
| GenBank mRNA | AB292648.1 | - | 2 | 213 | ||
| AF348451.1 | - | 2 | 103 | |||
| AK292754.1 | - | 2 | 233 | |||
| AK298400.1 | - | 2 | 98 | |||
| AK298420.1 | - | 2 | 228 | |||
| AK315331.1 | - | 2 | 260 | |||
| AK316495.1 | - | 2 | 98 | |||
| BC132819.1 | - | 2 | 309 | 40146841 | ||
| D13368.1 | - | 2 | 299 | |||
| X53414.1 | - | 2 | 313 | |||
| X56092.1 | - | 2 | 212 | |||

AGXT
Genomic Map