siRNA Details
| Gene Symbol: FUT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000149.3 | NP_000140.1 | 3 | 2342 | ||
| NM_001097639.1 | NP_001091108.1 | 3 | 1994 | |||
| NM_001097640.1 | NP_001091109.1 | 3 | 1940 | |||
| NM_001097641.1 | NP_001091110.1 | 2 | 1801 | |||
| XM_011527865.1 | XP_011526167.1 | 3 | 2320 | |||
| XM_011527866.1 | XP_011526168.1 | 3 | 1981 | |||
| XM_011527867.1 | XP_011526169.1 | 3 | 1950 | |||
| GenBank mRNA | AF131913.1 | - | 2 | 1873 | ||
| U27326.1 | - | 3 | 1933 | |||
| U27327.1 | - | 3 | 1987 | |||
| U27328.1 | - | 4 | 2336 | |||
| X53578.1 | - | 2 | 1776 | |||

FUT3
Genomic Map