siRNA Details
| Gene Symbol: OAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000274.3 | NP_000265.1 | 6 | 871 | ||
| NM_001171814.1 | NP_001165285.1 | 5 | 643 | |||
| NM_001322965.1 | NP_001309894.1 | 6 | 866 | |||
| NM_001322966.1 | NP_001309895.1 | 7 | 1186 | |||
| NM_001322967.1 | NP_001309896.1 | 7 | 991 | |||
| NM_001322968.1 | NP_001309897.1 | 8 | 1078 | |||
| NM_001322969.1 | NP_001309898.1 | 7 | 958 | |||
| NM_001322970.1 | NP_001309899.1 | 8 | 1174 | |||
| NM_001322971.1 | NP_001309900.1 | 4 | 550 | |||
| NM_001322974.1 | NP_001309903.1 | 6 | 757 | |||
| XM_017016279.1 | XP_016871768.1 | 6 | 2666 | |||
| GenBank mRNA | AB208817.1 | - | 6 | 2656 | ||
| AK296032.1 | - | 5 | 590 | |||
| AK312561.1 | - | 6 | 818 | |||
| AK315947.1 | - | 5 | 590 | |||
| BC000964.2 | - | 6 | 799 | 3449883 | ||
| BC016928.1 | - | 6 | 803 | 3913352 | ||
| CD683561.1 | - | 4 | 519 | |||
| CR457045.1 | - | 5 | 738 | |||
| CR749808.1 | - | 5 | 593 | |||
| M12267.1 | - | 6 | 792 | |||
| M14963.1 | - | 5 | 782 | |||
| M23204.1 | - | 6 | 789 | |||
| Y07511.1 | - | 5 | 778 | |||

OAT
Genomic Map