siRNA Details
| Gene Symbol: TPMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000367.3 | NP_000358.1 | 4 | 523 | ||
| XM_011514839.2 | XP_011513141.1 | 4 | 481 | |||
| XM_017011241.1 | XP_016866730.1 | 5 | 817 | |||
| GenBank mRNA | AF021876.1 | - | 3 | 346 | ||
| AF021877.1 | - | 3 | 346 | |||
| AK310420.1 | - | 4 | 417 | |||
| BC005339.1 | - | 4 | 433 | 3829338 | ||
| BC009596.1 | - | 4 | 432 | 4133029 | ||
| BI598325.1 | - | 4 | 508 | |||
| S62904.1 | - | 3 | 411 | |||
| U12387.1 | - | 4 | 422 | |||

TPMT
Genomic Map