siRNA Details
Gene Symbol: TPMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000367.3 | NP_000358.1 | 9 | 1206 | ||
XM_011514839.2 | XP_011513141.1 | 8 | 1119 | |||
XM_017011241.1 | XP_016866730.1 | 10 | 1500 | |||
GenBank mRNA | BC005339.1 | - | 9 | 1116 | 3829338 | |
BC009596.1 | - | 9 | 1115 | 4133029 | ||
S62904.1 | - | 8 | 1094 | |||
U12387.1 | - | 9 | 1105 |
