siRNA Details
| Gene Symbol: ULK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003565.2 | NP_003556.1 | 10, 11 | 1068 | ||
| XM_011538798.2 | XP_011537100.1 | 10, 11 | 845 | |||
| XM_011538799.2 | XP_011537101.1 | 10, 11 | 845 | |||
| GenBank mRNA | AB018265.2 | - | 10, 11 | 850 | ||
| AF045458.1 | - | 10, 11 | 1068 | |||

ULK1
Genomic Map