siRNA Details
| Gene Symbol: EPHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017000534.1 | XP_016856023.1 | 17 | 3628 | ||
| XM_017000535.1 | XP_016856024.1 | 17 | 3588 | |||
| XM_017000536.1 | XP_016856025.1 | 16 | 3436 | |||
| GenBank mRNA | BC008655.1 | - | 5 | 1190 | ||
| BC037166.2 | - | 17 | 3476 | 5090654 | ||
| M59371.1 | - | 17 | 3471 | |||

EPHA2
Genomic Map