siRNA Details
| Gene Symbol: DAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014326.3 | NP_055141.2 | 12 | 1588 | ||
| XM_011521413.2 | XP_011519715.1 | 11 | 1786 | |||
| XM_011521414.1 | XP_011519716.1 | 11 | 4179 | |||
| XM_011521417.1 | XP_011519719.1 | 10 | 1517 | |||
| XM_011521421.2 | XP_011519723.1 | 10 | 1596 | |||
| XM_017022044.1 | XP_016877533.1 | 11 | 4175 | |||
| XM_017022045.1 | XP_016877534.1 | 10 | 1740 | |||
| GenBank mRNA | AB018001.1 | - | 12 | 1587 | ||
| AF052941.1 | - | 12 | 1588 | |||
| AK026801.1 | - | 1 | 1130 | |||

DAPK2
Genomic Map