siRNA Details
| Gene Symbol: PASK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252119.1 | NP_001239048.1 | 3 | 537 | ||
| NM_001252120.1 | NP_001239049.1 | 4 | 690 | |||
| NM_001252122.1 | NP_001239051.1 | 3 | 537 | |||
| NM_001252124.1 | NP_001239053.1 | 3 | 537 | |||
| NM_015148.3 | NP_055963.2 | 3 | 537 | |||
| XM_011510826.1 | XP_011509128.1 | 3 | 492 | |||
| XM_011510828.1 | XP_011509130.1 | 4 | 593 | |||
| XM_011510829.1 | XP_011509131.1 | 3 | 534 | |||
| XM_011510830.1 | XP_011509132.1 | 3 | 535 | |||
| XM_017003634.1 | XP_016859123.1 | 3 | 781 | |||
| XM_017003635.1 | XP_016859124.1 | 4 | 593 | |||
| XM_017003636.1 | XP_016859125.1 | 3 | 549 | |||
| GenBank mRNA | AF387103.1 | - | 3 | 477 | ||
| AK056516.1 | - | 3 | 482 | |||
| AK302417.1 | - | 3 | 493 | |||
| BC050565.1 | - | 4 | 496 | 4537404 | ||
| BC063585.1 | - | 3 | 480 | 4419998 | ||
| CR749231.1 | - | 4 | 691 | |||
| D50925.2 | - | 3 | 468 | |||
| DA104684.1 | - | 3 | 538 | |||

PASK
Genomic Map