siRNA Details
| Gene Symbol: COQ8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020247.4 | NP_064632.2 | 3 | 662 | ||
| XM_005273201.1 | XP_005273258.1 | 3 | 558 | |||
| XM_011544238.1 | XP_011542540.1 | 4 | 754 | |||
| XM_011544239.2 | XP_011542541.1 | 5 | 777 | |||
| XM_011544240.2 | XP_011542542.1 | 3 | 571 | |||
| XM_011544241.2 | XP_011542543.1 | 3 | 533 | |||
| XM_017001852.1 | XP_016857341.1 | 3 | 752 | |||
| GenBank mRNA | AB073905.1 | - | 3 | 585 | ||
| AK074693.1 | - | 3 | 605 | |||
| AK090494.1 | - | 3 | 399 | |||
| AK092784.1 | - | 3 | 385 | |||
| AK293571.1 | - | 4 | 559 | |||
| AK301012.1 | - | 5 | 583 | |||
| BC005171.2 | - | 3 | 611 | 3604948 | ||
| BC129930.1 | - | 9 | 1504 | 40010265 | ||
| BX648860.1 | - | 3 | 483 | |||

COQ8A
Genomic Map