siRNA Details
| Gene Symbol: CERK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022766.5 | NP_073603.2 | 13 | 2134 | ||
| XM_017028909.1 | XP_016884398.1 | 13 | 1931 | |||
| GenBank mRNA | AB051433.1 | - | 12 | 1860 | ||
| AB079066.1 | - | 13 | 2150 | |||
| AK026892.1 | - | 1 | 292 | |||
| AL832794.1 | - | 3 | 711 | |||
| BC004278.1 | - | 3 | 711 | |||
| BC108712.1 | - | 2 | 636 | |||

CERK
Genomic Map