siRNA Details
| Gene Symbol: ITGAE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002208.4 | NP_002199.3 | 3 | 329 | ||
| XM_011523823.2 | XP_011522125.1 | 3 | 767 | |||
| XM_011523825.2 | XP_011522127.1 | 3 | 765 | |||
| XM_011523827.2 | XP_011522129.1 | 3 | 759 | |||
| XM_011523828.2 | XP_011522130.1 | 3 | 763 | |||
| XM_017024586.1 | XP_016880075.1 | 4 | 2152 | |||
| XM_017024587.1 | XP_016880076.1 | 4 | 2152 | |||
| GenBank mRNA | BC113436.1 | - | 3 | 319 | 8322488 | |
| BC117207.1 | - | 3 | 319 | 40125758 | ||
| L25851.2 | - | 3 | 355 | |||

ITGAE
Genomic Map