siRNA Details
Gene Symbol: DLSTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130749.1 | 1 | 961 |
Gene Symbol: DLST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001933.4 | NP_001924.2 | 10 | 788 | ||
NR_033814.1 | 9 | 730 | ||||
NR_045209.1 | 9 | 739 | ||||
GenBank mRNA | AK289414.1 | - | 10 | 786 | ||
AK299505.1 | - | 7 | 622 | |||
AK316463.1 | - | 9 | 705 | |||
AL538322.3 | - | 3 | 228 | |||
BC000302.2 | - | 10 | 750 | 2820348 | ||
BC001922.1 | - | 10 | 750 | 2820348 | ||
BX248276.1 | - | 9 | 707 | |||
BX248764.1 | - | 8 | 619 | |||
BX248774.1 | - | 9 | 674 | |||
CR456727.1 | - | 10 | 725 | |||
D16373.1 | - | 10 | 737 | |||
D17297.1 | - | 1 | 2088 | |||
L37418.1 | - | 10 | 725 |
