siRNA Details
| Gene Symbol: DLST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001933.4 | NP_001924.2 | 15 | 2073 | ||
| NR_033814.1 | 14 | 2015 | ||||
| NR_045209.1 | 14 | 2024 | ||||
| GenBank mRNA | BC000302.2 | - | 15 | 2035 | 2820348 | |
| BC001922.1 | - | 15 | 2035 | 2820348 | ||
| CN422511.1 | - | 1 | 96 | |||

DLST
Genomic Map