siRNA Details
| Gene Symbol: KLRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002258.2 | NP_002249.1 | 5 | 499 | ||
| GenBank mRNA | AK292022.1 | - | 5 | 514 | ||
| BC113997.1 | - | 5 | 487 | |||
| BC114516.1 | - | 5 | 487 | 40031215 | ||
| BF432813.1 | - | 5 | 500 | |||
| U11276.1 | - | 5 | 497 | |||

KLRB1
Genomic Map