siRNA Details
| Gene Symbol: LEPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003679.3 | NP_001003679.1 | 4 | 311 | ||
| NM_001003680.3 | NP_001003680.1 | 4 | 311 | |||
| NM_001198687.1 | NP_001185616.1 | 3 | 276 | |||
| NM_001198688.1 | NP_001185617.1 | 3 | 276 | |||
| NM_001198689.1 | NP_001185618.1 | 3 | 276 | |||
| NM_002303.5 | NP_002294.2 | 4 | 311 | |||
| GenBank mRNA | AK290208.1 | - | 4 | 276 | ||
| BC031086.1 | - | 3 | 276 | |||
| BC131779.1 | - | 4 | 243 | |||
| U43168.1 | - | 4 | 319 | |||
| U50748.1 | - | 2 | 126 | |||
| U52912.1 | - | 3 | 222 | |||
| U52913.1 | - | 3 | 222 | |||
| U52914.1 | - | 3 | 222 | |||
| U66495.1 | - | 4 | 319 | |||
| U66496.1 | - | 4 | 319 | |||
| U66497.1 | - | 3 | 252 | |||

LEPR
Genomic Map