siRNA Details
| Gene Symbol: PKN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002741.3 | NP_002732.3 | 17 | 2372 | ||
| NM_213560.1 | NP_998725.1 | 17 | 2239 | |||
| XM_011528128.1 | XP_011526430.1 | 17 | 2255 | |||
| GenBank mRNA | AK123007.1 | - | 17 | 2253 | ||
| AK292130.1 | - | 17 | 2236 | |||
| AK313886.1 | - | 17 | 2212 | |||
| BC018575.1 | - | 11 | 1090 | |||
| BC040061.1 | - | 17 | 2369 | 5752583 | ||
| BC094766.1 | - | 15 | 2713 | 30342125 | ||
| D26181.1 | - | 17 | 2240 | |||
| S75546.1 | - | 17 | 2288 | |||
| U33053.1 | - | 17 | 2288 | |||

PKN1
Genomic Map