siRNA Details
| Gene Symbol: GRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002929.2 | NP_002920.1 | 1 | 335 | ||
| GenBank mRNA | AK289912.1 | - | 1 | 459 | ||
| BC140945.1 | - | 1 | 300 | 9056757 | ||
| L77503.1 | - | 1 | 242 | |||
| U63973.1 | - | 1 | 335 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRK1| Gene Symbol: GRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002929.2 | NP_002920.1 | 1 | 335 | ||
| GenBank mRNA | AK289912.1 | - | 1 | 459 | ||
| BC140945.1 | - | 1 | 300 | 9056757 | ||
| L77503.1 | - | 1 | 242 | |||
| U63973.1 | - | 1 | 335 | |||